AutoLys M Tube Pliers
Applied Biosystems™
AutoLys M Tube Pliers
AutoLys M Tube Pliers can be used to separate the inner-tube from the outer-tube of AutoLys M Tubes. Centrifugation ofRead more
Have Questions?
Catalog NumberQuantity
A382611 each
Catalog number A38261
Price (USD)
80.65
Online Exclusive
86.25
Save 5.60 (6%)
Each
Quantity:
1 each
Price (USD)
80.65
Online Exclusive
86.25
Save 5.60 (6%)
Each
AutoLys M Tube Pliers can be used to separate the inner-tube from the outer-tube of AutoLys M Tubes. Centrifugation of the tubes in an AutoLys M Tube Rack is required when processing FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit. The outer-tube will contain the lysate that is processed further to recover the nucleic acids.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AutoLys TubeLifter System
Quantity1 each
Shipping ConditionRoom Temperature
Unit SizeEach
Frequently asked questions (FAQs)
Can I use a heat block for the AutoLys M tube incubation?
No. The AutoLys M tubes don't fit in a standard 1.5 mL or 2.0 mL heat block. We recommend incubating the tubes in the tube racks in an incubator or oven.
What kind of centrifuge do I need to spin down the AutoLys M Tubes (Cat. No. A38738)?
Any centrifuge with plate adapters will work. The AutoLys M Tube Racks fit nicely into those adapters.
Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?
No, the AutoLys M Tubes can be lifted manually, but the tools significantly reduce hand strain and increase ease of use as well as throughput.
Do the AutoLys M Tubes come with the MagMAX FFPE DNA/RNA Ultra Kit?
No, they have to be purchased separately together with some other products that are necessary for using the AutoLys M Tube system efficiently:
- AutoLys M Tubes and Caps (Cat. No. A38738)
- Autolys M Tube Locking Lid (Cat. No. A37954)
- AutoLys M TubeLifter (Cat. No. A37956)
- AutoLys M Tube Pliers (Cat. No. A38261)
- AutoLys M Tube Racks (Cat. No. A37955)
How does using the the AutoLys M Tube system help in increase the DNA/RNA yield from FFPE samples?
The FFPE-associated wax and debris are held in the upper chamber of the AutoLys M tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate. The AutoLys M tubes thus reduce manipulation of the sample eliminating the need to pellet the tissue and wash, so tissue loss is minimized