Countess™ Cell Counting Chamber Slides
Invitrogen™
Countess™ Cell Counting Chamber Slides
Countess Cell Counting Chamber Slides are conveniently packaged for use with the Countess Automated Cell Counters. Each disposable and reusable slide features two enclosed chambers.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoTypeIncludesQuantity
A40008469Cell Counting Chamber SlideSafeCount Stain, 2 x 1 mL Vials50 Slides
A25750Reusable SlideNo Stain1 Glass Slide
C10283Cell Counting Chamber SlideNo Stain50 Slides
C10228Cell Counting Chamber SlideTrypan Blue Stain, 2 x 1 mL Vials50 Slides
C10312Cell Counting Chamber SlideTrypan Blue Stain, 20 x 1 mL Vials500 Slides
C10313Cell Counting Chamber SlideTrypan Blue Stain, 100 mL Bottle1250 Slides
C10314Cell Counting Chamber SlideTrypan Blue Stain, 100 mL Bottle2500 Slides
C10315Cell Counting Chamber SlideTrypan Blue Stain, 2 x 100 mL Bottles5000 Slides
Número do catálogo A40008469
Preço (BRL)
-
Type:
Cell Counting Chamber Slide
Includes:
SafeCount Stain, 2 x 1 mL Vials
Quantity:
50 Slides
The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.
The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.
The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.
Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.
The Countess Automated Cell Counter works in three simple steps:
  • Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
  • Insert the slide into the instrument
  • Press 'Count'
That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Coupler OptionDead Cell Stain
For Use With (Equipment)Countess 3/3FL Automated Cell Counters
IncludesSafeCount Stain, 2 x 1 mL Vials
Quantity50 Slides
TypeCell Counting Chamber Slide
Unit SizeEach
Conteúdo e armazenamento
50 Pk. Chamber Slides and SafeCount (Cat. No. A40008024), Store at Room Temp