Mass Spectrometry Grade Proteases
Thermo Scientific™

Mass Spectrometry Grade Proteases

Improve sequence coverage and protein identification for mass spectrometry with high-quality, MS-grade proteases provided in stable liquid or lyophilized formats. Suitable for either in-solution or in-gel digestion workflows, each protease may be used individually or in combination to improve protein sequence coverage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
A40009100 μgTrypsin/Lys-C Protease Mix
900532 μgAsp-N Endoproteinase
900564 x 25 μgChymotrypsin Endoproteinase
900545 x 10 μgGlu-C Endoproteinase
903051 x 100 μgTrypsin Protease
900591 mgTrypsin Protease
A4000720 μgTrypsin/Lys-C Protease Mix
A410075 x 20 μgTrypsin/Lys-C Protease Mix
900511 x 20 μgLys-C Endoproteinase
90307100 μgLys-C Endoproteinase
900575 x 20 μgTrypsin Protease
900585 x 100 μgTrypsin Protease
Catalog number A40009
Price (USD)
186.65
Online exclusive
212.00
Save 25.35
Each
Quantity:
100 μg
Product Type:
Trypsin/Lys-C Protease Mix
Price (USD)
186.65
Online exclusive
212.00
Save 25.35
Each
Protein characterization, identification, and quantification by MS begins with efficient, reproducible protein digestion. Sequence specific proteases are utilized to cleave proteins into smaller fragments or peptides. Trypsin is the protease of choice for protein digestion. However, separate or sequential digestion with alternative proteases can improve individual protein sequence coverage, or generate unique peptide sequences for different MS applications.
  • Exceptional selectivity—trypsin has >95% C-terminal lysine and arginine specificity; LysC has >90% C-terminal lysine cleavage specificity
  • High purity—no detectable chymotrypsin activity (trypsin)
  • Complete digestion—trypsin/LysC enzyme combination reduces tryptic missed cleavages
  • Enhanced stability—trypsin chemically modified for reduced autolytic activity
  • Convenient—enzymes provided in stable lyophilized or liquid formats

Trypsin Protease is a mass spectrometry (MS)-grade serine protease derived from porcine pancreatic extracts that specifically cleaves at the carboxyl side of lysine and arginine residues. The enzyme is TPCK-treated to eliminate chymotryptic activity and chemically modified by methylation, yielding a highly active and more stable form of the enzyme. Trypsin Protease can tolerate commonly used partially denaturing conditions, such as 0.1% SDS, 1 M urea, and 10% acetonitrile. Trypsin Protease is most active at pH 7 to 9 and can be reversibly inactivated at pH < 4. Trypsin can be purchased lyophilized in 1-mg 5-packs of 20 μg or 100 μg quantities and in solution at 100 μg.

Lys-C Protease is a mass spectrometry (MS)-grade serine protease isolated from Lysobacter enzymogenes. Lys-C Protease has high activity and specificity for lysine residues, resulting in larger peptides and less sample complexity than trypsin (i.e., fewer peptides). Unlike trypsin, Lys-C Protease can cleave lysines followed by prolines, making it ideal for sequential protein digestion followed by trypsin to decrease missed cleavages. These unique Lys-C Protease properties ensure high digestion efficiency when used alone or followed by tryptic digestion. Additionally, Lys-C prototypic peptides typically have higher charge states, making it an enzyme of choice for use with ETD fragmentation.

Lys-C Protease is commonly used in phosphopeptide enrichment workflows because it generates peptides with primary amines at both the N- and C-terminus, allowing the fragments to be double-labeled with amine-reactive isobaric tags. This results in enhanced peptide ionization and improved limits of quantitation since more fragment ions can be re-isolated during MS3 acquisition. This enzyme can be used for in-solution or in-gel digestion workflows to produce peptides for LC-MS/MS protein identification.

Efficient protein digestion can be completed in two hours at 37°C. Lys-C Protease remains active in highly denaturing conditions, such as 8 M urea, 2 M guanidine-HCl, 1% SDS, 2% CHAPS, and 40% acetonitrile, and functions well at pH 7–9 (maximal activity at pH 8). This lyophilized enzyme has a mass of 30 kDa and is stable for one year when stored at –20°C. This Lys-C enzyme is packaged lyophilized (20 μg or 100 μg quantities).

Trypsin/Lys-C Protease Mix is a lyophilized mixture of trypsin and LysC proteases that has been optimized to improve digestion efficiency of proteins. Although trypsin is routinely used for protein digestion, this protease alone is not sufficient to fully digest proteins at the carboxyl-end of lysine and arginine residues. Therefore, Lys-C protease is commonly combined with trypsin to sequentially digest proteins with fewer missed cleavages. Trypsin/Lys-C Protease Mix is provided in flexible formats of 20 μg, 5 x 20 μg, or 100 μg. Digestion can be completed in as little as 1.5–3 hours or up to overnight, depending on enzyme to protein ratio.

Chymotrypsin, GluC, Asp-N Proteases

  • Increased sequence coverage—better protein characterization results from overlapping peptides with complementary chromatographic, ionization, and fragmentation properties
  • High specific activity—each protease has excellent enzymatic specificity
  • Stable—provided in a lyophilized format

Asp-N Protease is an MS-grade zinc metalloproteinase derived from a mutant strain of Pseudomonas fragi and requires a trace amount of zinc for activity. Asp-N Protease cleaves primarily at the amino side of aspartate and cysteic acid that results from cysteine oxidation. Cleavage can also occur at glutamic acid; however, the rate of cleavage at the glutamyl residue is significantly lower than the rate of cleavage at the aspartic acid residue. Asp-N Protease can efficiently digest protein in 2–20 hours at 37°C with activity greater than 20,000 units/mg protein and remains active under denaturing conditions, such as 1 M urea, 2 M guanidine·HCl, 0.1% SDS, 2% CHAPS, and 10% acetonitrile, with optimal activity at pH 6–8. This lyophilized enzyme has a mass of 27 kDa and is stable for one year when stored at -20°C.

Glu-C Protease, also referred to as V-8 protease, is an MS-grade serine protease isolated from Staphylococcus aureus. Glu-C Protease specifically cleaves the carboxyl side of glutamic acid residues in ammonium bicarbonate and ammonium acetate buffers, generating a limited number of peptide fragments. Cleavage can also occur at both glutamic and aspartic acid residues in phosphate buffers. Glu-C Protease can efficiently digest protein in 5–18 hours at 37°C with activity greater than 500 units/mg protein and remains active under denaturing conditions, such as 2 M urea, 1 M guanidine·HCl, 0.1% SDS, 2% CHAPS, and 20% acetonitrile. Glu-C Protease activity is optimal at pH 8. This lyophilized enzyme has a mass of 27 kDa and is stable for one year when stored at -20°C.

Chymotrypsin Protease is an MS-grade endoproteinase isolated from bovine pancreas that specifically cleaves at the carboxyl side of tyrosine, phenylalanine, tryptophan, and leucine. Two predominant forms of chymotrypsin protease, A and B, are found in equal amounts in bovine pancreas. They are similar proteins (80% homology) but have different proteolytic characteristics. Both forms of chymotrypsin protease are present in Thermo Scientific Chymotrypsin Protease. Since trypsin may co-purify with chymotrypsin protease derived from natural sources, Chymotrypsin Protease has been treated with TLCK to eliminate potential tryptic activity, improving digestion specificity. Chymotrypsin Protease can tolerate mild denaturing conditions, such as 0.1% SDS, 2 M urea, 2 M guanidine·HCl, 1% CHAPS, and 30% acetonitrile, with optimal activity at pH 7.5–8.5. This lyophilized enzyme has a mass of 25 kDa and is stable for one year when stored at -20°C.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypePeptides
For Use With (Equipment)Mass Spectrometer
GradeMS
Quantity100 μg
Shipping ConditionWet Ice
Workflow StepProtein Digestion
Detection MethodMass Spectrometry
FormSolid
Product LinePierce
Product TypeTrypsin/Lys-C Protease Mix
Starting MaterialProtein Samples
Unit SizeEach
Contents & Storage
Store at -20°C

Frequently asked questions (FAQs)

What is the best enzyme to use for protein digestion for mass spectrometry samples?

Trypsin (Cat. Nos. 90057, 90058) or Trypsin/LysC mix (Cat. Nos. A40007, A40009, A41007) are most commonly used for proteomic applications in order to ensure reproducibility and complete digestion. Other commonly used enzymes for purified protein characterization and unique applications include Chymotrypsin Protease, MS Grade (Cat. No. 90056), Immobilized Pepsin (Cat. No. 20343), LysN Protease, MS Grade (Cat. No. 90300), Asp-N Protease, MS Grade (Cat. No. 90053), and Glu-C Protease, MS Grade (Cat. No. 90054).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Is the trypsin in Pierce Trypsin/Lys-C Protease Mix, MS-Grade modified?

Yes, the MS-Grade trypsin protease in this mix is derived from porcine pancreatic extracts and has been Tosyl phenylalanyl chloromethyl ketone (TPCK)-treated to eliminate chymotryptic activity, and methylated to improve stability during digestion.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why are trypsin and Lys-C used together in the Pierce Trypsin/Lys-C Protease Mix, MS-Grade?

Trypsin cuts at the carboxyl end of lysine and arginine residues, and Lys-C protease cuts at the carboxyl end of just lysine residues with Lys-C being more efficient than trypsin. Although trypsin is routinely used for protein digestion, this protease alone is not sufficient to fully digest proteins at the carboxyl-end of lysine and arginine residues. Therefore, Lys-C is combined with trypsin to sequentially digest proteins with fewer missed cleavage points at lysines.

The added benefit of having trypsin and Lys-C together is that unlike trypsin, Lys-C can cleave lysines followed by prolines.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (3)

Citations & References
Abstract
Integrating Vascular Phenotypic and Proteomic Analysis in an Open Microfluidic Platform.
Authors:Jung S,Cheong S,Lee Y,Lee J,Lee J,Kwon MS,Oh YS,Kim T,Ha S,Kim SJ,Jo DH,Ko J,Jeon NL
Journal:ACS nano
PubMed ID:39208278
This research introduces a vascular phenotypic and proteomic analysis (VPT) platform designed to perform high-throughput experiments on vascular development. The VPT platform utilizes an open-channel configuration that facilitates angiogenesis by precise alignment of endothelial cells, allowing for a 3D morphological examination and protein analysis. We study the effects of antiangiogenic ... More
Sex differences in single neuron function and proteomics profiles examined by patch-clamp and mass spectrometry in the locus coeruleus of the adult mouse.
Authors:Lee J,Wang ZM,Messi ML,Milligan C,Furdui CM,Delbono O
Journal:Acta physiologica (Oxford, England)
PubMed ID:38459766
AIMS: This study aimed to characterize the properties of locus coeruleus (LC) noradrenergic neurons in male and female mice. We also sought to investigate sex-specific differences in membrane properties, action potential generation, and protein expression profiles to understand the mechanisms underlying neuronal excitability variations. METHODS: Utilizing a genetic mouse model ... More
Discovery of electrophilic degraders that exploit S(N)Ar chemistry.
Authors:Zhuang Z,Byun WS,Kozicka Z,Dwyer BG,Donovan KA,Jiang Z,Jones HM,Abeja DM,Nix MN,Zhong J,Słabicki M,Fischer ES,Ebert BL,Gray NS
Journal:bioRxiv : the preprint server for biology
PubMed ID:39386645
Targeted covalent inhibition (TCI) and targeted protein degradation (TPD) have proven effective in pharmacologically addressing formerly 'undruggable' targets. Integration of both methodologies has resulted in the development of electrophilic degraders where recruitment of a suitable E3 ubiquitin ligase is achieved through formation of a covalent bond with a cysteine nucleophile. ... More