SeqStudio™ Cartridge v2
Applied Biosystems™

SeqStudio™ Cartridge v2

The Applied Biosystems SeqStudio Cartridge v2 is an all-in-one and easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System.
Have Questions?
Catalog NumberQuantity
A41331Up to 250 Injections or 1000 Reactions
Catalog number A41331
Price (USD)
2,797.65
Online exclusive
2,945.00
Save 147.35
Each
Quantity:
Up to 250 Injections or 1000 Reactions
Price (USD)
2,797.65
Online exclusive
2,945.00
Save 147.35
Each

The Applied Biosystems SeqStudio Cartridge v2 is an all-in-one and easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System. The universal polymer allows for flexibility to perform Sanger sequencing and fragment analysis on the same instrument run with the same cartridge. Just load your samples, click in the cartridge, and go.

Key features:

  • Minimize hands-on setup time with all-in-one cartridge
  • Track consumables usage through radio frequency identification (RFID)
  • Extended on-instrument 6-month shelf life for cartridge after opening
  • Allows for 250 injections or 1000 reactions per cartridge*
  • Responsible, simple, no-charge recycling program

*SeqStudio Cartridge v2 is optimized to work with the latest software version for the Seqstudio Genetic Analyzer. Please ensure that your Seqstudio Genetic Analyzer is running the latest software before using this cartridge.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Capillary Length28 cm
For Use With (Application)Rare Variant Sequencing, Resequencing, Comparative Sequencing (Somatic Mutations - 10:50), Fragment Analysis (Denaturing), Short Read Sequencing, De Novo Sequencing - Low to Mid Throughput, Short PCR Product Sequencing
Format96-well Plate, 8-tube Strip
Height (Metric)44.2 cm
Length (Metric)49.5 cm
Product TypeCartridge V2
QuantityUp to 250 Injections or 1000 Reactions
Shipping ConditionWet Ice
Throughput4 Capillary Instrument
Width (Metric)64.8 cm
Product LineSeqStudio
Unit SizeEach
Contents & Storage
Sufficient for 250 injections/1000 reactions

Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

What is included in the SeqStudio Cartridge v2 (Cat. No. A41331) and can the cartridge (Cat. No. A41328) be purchased separately?

The SeqStudio Cartridge v2 includes the SeqStudio Genetic Analyzer Cartridge v2 and a SeqStudio Integrated Capillary Protector (Cat. No. A31923). Even though the cartridge has "A41328" on the box, it cannot be purchased separately and is only available as part of the SeqStudio Cartridge v2.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

How do I recycle my SeqStudio cartridge if my country is not supported by the recycling program?

We would recommend disposing of your SeqStudio cartridge according to your country's regulations on proper disposal of laboratory equipment and reagents.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

How do I know if the SeqStudio cartridge recycling program is supported in my country?

You can view the details and availability of the SeqStudio Cartridge recycling program on this web page (https://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/sanger-sequencing-technology-accessories/seqstudio-genetic-analyzer/seqstudio-cartridge-recycling-program.html) or speak to your local representative for more information.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Where does the recycling of my used SeqStudio cartridge take place?

We have partnered with E-End Solutions in North America and SIMMs Recycling Solutions in Europe for recycling of used SeqStudio cartridges. More details about the program can be found in the SeqStudio Cartridge Green Fact Sheet (https://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/seqstudio-cartridge-green-fact-sheet.pdf).

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.