Product Image
Ion Torrent™
Ion Chef™ Instrument for Agrigenomics
The Ion Chef System is the next generation of workflow simplification products for the Ion GeneStudio S5 Systems. The Ion자세히 알아보기
Have Questions?
카탈로그 번호수량
A435581 instrument
카탈로그 번호 A43558
제품 가격(KRW)
-
수량:
1 instrument
The Ion Chef System is the next generation of workflow simplification products for the Ion GeneStudio S5 Systems. The Ion Chef System provides automated library preparation, template preparation, and chip loading for users at any experience level. In less than 15 minutes of up-front hands-on time and with the use of pre-packaged library preparation reagent kits, the Ion Chef System provides a convenient walk-away workflow resulting in equalized, pooled libraries ready for templating. The automated workflow supports Ion AmpliSeq one- and two-pool designs. Visit www.AmpliSeq.com to design your own custom panels. After library preparation, less than 15 minutes of hands-on time is required to set up the Ion Chef System to perform fully automated template preparation, producing one or two chips ready for sequencing. Automating your sample preparation workflows on the Ion Chef System results in significantly higher productivity.
The Ion Chef System:
• Simplifies the workflow
• Minimizes sources of user-introduced variability
• Helps save time and labor
• Supports sequencing preparation for the Ion GeneStudio S5 Systems
The Ion Chef System features:
• Automated Ion Ampliseq library preparation, template preparation, and chip loading
• Cartridge-based reagent system
• Sample and reagent-tracking capability
• Intuitive user interface
• On-board diagnostics
Simplify the Ion Torrent NGS workflow
The Ion Chef System simplifies the Ion Torrent next-generation sequencing (NGS) workflow by integrating several manual and instrument steps into a single process. The Ion Chef System workflow now incorporates all steps of library preparation (targeted PCR amplification with respective primer panel, partial digestion of primer sequences, adapter/barcode ligation, purification, library equalization and pooling), as well as all steps of template preparation and chip loading. The simple, automated workflow provides easy access for users of any experience level.
Minimize variability and load chips reproducibly
The Ion Chef System provides reproducible chip loading by automating multiple manual chip-loading steps. The sophisticated visual system inside the Ion Chef System is designed to help reduce user-introduced error and variability by providing barcode reading and positive identification of samples, chips, and reagents. The visual system performs the setup check before the run, as well as the reagents and consumables load and unload check, calibration, and diagnostics.
Save time and labor
The Ion Chef System enables significant time and labor savings by offering quick and easy setup for both library preparation and templating that typically requires less than 15 minutes of hands-on time for each step.
Support for all Ion Torrent sequencing workflows
The flexible Ion Chef System is designed to provide automated and consistent Ion AmpliSeq library preparation and template preparation for sequencing on the Ion GeneStudio S5 Systems using any of the corresponding chips or template preparation and sequencing chemistries.
Services and support
Service and support plans—comprehensive plans to maximize instrument performance and ensure availability of critical systems
Qualification services—document and ensure instrument is installed, operating, and performing to manufacturer’s specifications
Training—courses for experimental design, data analysis, troubleshooting, applications, and instrument operation
Bioinformatics—tailored support for NGS informatics execution, software use, data analysis, and downstream applications
DataSafe Solution—storage and back-up strategy, design, hardware, and support for all lab data
For Research Use Only. Not for use in diagnostic procedures.
사양
제품 유형Chef Instrument
수량1 instrument
배송 조건Room Temperature
제품라인Ion Chef
Unit SizeEach
구성 및 보관
Store at room temperature.