Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
Product Image
Ion Torrent™
Ion 550™ Single Chip Supplemental Kit for Agrigenomics
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required for use with theWeitere Informationen
Have Questions?
KatalognummerMenge
A435594 Reaktionen
Katalognummer A43559
Preis (EUR)
-
Menge:
4 Reaktionen
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required for use with the single-chip Ion Chef workflow. The Ion 550 Single Chip Supplemental Kit, together with the Ion 550 Kit—Chef, enables the flexibility to prepare one loaded chip per Ion Chef run, without loss of templating reagents.
Note: This supplemental templating kit for Ion Chef is only compatible with the Ion 550 Chip and the Ion 550 Kit—Chef.
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
Zur Verwendung mit (Geräte)Ion 550 Chip, Ion 550 Kit-Chef
ProdukttypSingle Chip Supplemental Kit
Menge4 Reaktionen
VersandbedingungZugelassen für den Versand bei Raumtemperatur oder auf nassem Eis
ProduktlinieIon Torrent
Unit SizeEach
Inhalt und Lagerung
• 4 Ion S5 Chef Supplies, store at room temperature
• 1 Ion S5 Sequencing Solutions, store at room temperature
• 1 Ion S5 Sequencing Reagents, store at -20°C
Häufig gestellte Fragen (FAQ)
Do I need to purchase the Ion 550 Single Chip Supplemental Kit?
The Ion 550 Kit-Chef contains Ion Chef templating and sequencing consumables sufficient for preparing and sequencing eight Ion 550 chips for four Ion Chef runs and four sequencer initializations (dual chip run configuration). If you are running in the single chip configuration, then you will need to purchase the Ion 550 Single Chip Supplemental Kit separately to ensure that you have enough templating and sequencing consumables to support eight single chip Ion Chef templating runs and eight sequencer initializations with the Ion 550 Kit-Chef.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is in the Ion 550 Single Chip Supplemental Kit and how is it shipped/stored?
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required to support eight single chip Ion Chef runs and eight sequencer initializations with the Ion 550 Kit-Chef. The Ion 550 Single Chip Supplemental Kit includes the following components:
- Ion S5 Chef Supplies (4 boxes) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Solutions (1 box) (ships at room temperature; store at room temperature)
- Ion S5 Sequencing Reagents (1box) (ships on wet ice at 2-8 degrees C; store at -20 degrees C)
IMPORTANT! Do not store the Ion S5 Sequencing Reagents (Part No. A27768) on dry ice or in a closed environment containing dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.