Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Countess™ 3 USB Drive
Invitrogen™
Countess™ 3 USB Drive
Save and transfer cell counting data from the Countess 3 and 3 FL automated cell counters using the Countess 3 USB Drive. This 32 GB USB drive is included with the Countess 3 and 3 FL instruments or a replacement USB drive can be purchased separately for additional storage.
Have Questions?
Catalog NumberQuantity
A482091 drive
Catalog number A48209
Price (USD)
32.10
Each
Quantity:
1 drive
Price (USD)
32.10
Each
The Countess 3 USB Drive is included with both the Countess 3 and Countess 3 FL automated cell counters for saving and transferring cell counting data from the instruments. For additional storage and convenience, a replacement Countess 3 USB drive is offered here as a separate purchase. It is a 32 GB USB drive that is FAT32- and NTFS-formatted for use with Countess 3 and Countess 3 FL automated cell counters, as well as other USB-enabled devices.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CompatibilityFAT and NTFS formatting
For Use With (Equipment)Countess 3 and 3 FL Automated Cell Counters
Quantity1 drive
Product LineCountess
TypeUSB Drive
Unit SizeEach
Frequently asked questions (FAQs)
Where can I find the service retirement letter for the Countess II and II FL Automated Cell Counters?
You can find the service retirement letter for Countess II and II FL Automated Cell Counters below:
Important end-of-life communication for Countess II and II FL products
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Where can I find FAQs for Countess II and II FL Automated Cell Counters?
Countess II and II FL Automated Cell Counter FAQs can be found in our Product FAQ Database.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Where can I find a troubleshooting information for Countess 3 and 3 FL Automated Cell Counters?
Troubleshooting information for Countess 3 and 3FL can be found on the Cell Counting Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the shelf life for Countess Cell Counting Chamber Slides, disposable?
Our guarantee period for disposable Countess Cell Counting Chamber Slides is 5 years.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How can I find information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter?
For information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter, including 21 CFR Part 11, IQ/OQ, and warranty packages, please see refer to Countess Automated Cell Counter Services on our website.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.