Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Countess™ 3 Standard Slide
Invitrogen™
Countess™ 3 Standard Slide
The Countess 3 Standard Slide can be used as a reference or for demonstration of the cell counting, viability, and fluorescence features of the Countess automated cell counters. This ready-to-use and reusable slide contains permanently mounted microspheres that mimic the appearance of cell samples.
Have Questions?
Catalog NumberQuantity
A518761 slide
Catalog number A51876
Price (USD)
168.00
Each
Quantity:
1 slide
Price (USD)
168.00
Each
The Countess 3 Standard Slide is a ready-to-use and reusable slide used as a reference for cell counting, viability, and fluorescence on the Countess automated cell counters. This slide is optimized for ensuring accurate performance in brightfield cell counting applications when using Countess II, Countess II FL, Countess 3, and Countess 3 FL automated cell counters. The permanently mounted microspheres on the Countess 3 Standard Slide enable repeatable counts to be easily achieved over the life of the slide, with variability only encountered upon changing instrument gating settings.
The Countess 3 Standard Slide is supplied ready-to-use and does not require any special handling or sample preparation, thus eliminating the major source of count-to-count variability. This makes it an ideal reference slide that can be used to achieve consistent and repeatable cell counts and viability measurements across days or between Countess 3 instruments. The slide can also be used for demonstrating how to use the Countess automated cell counters for both brightfield and fluorescence cell counting.
Features of the Countess 3 Standard Slide include:
  • Accurate – ready-to-use slide helps eliminate the error involved in creating standard solutions and dilution series
  • Flexible – optimized for use in Countess II, Countess II FL, Countess 3, and Countess 3 FL automated cell counters
  • Fast – supplied as a pre-loaded, immobilized, bead suspension; no preparation or mixing required
  • Convenient – one standard slide for both brightfield and fluorescence cell counting applications
Immobilized microspheres mimic cell counting samples
The Countess 3 Standard Slide contains 15 μm microspheres that are permanently mounted in a cell counting chamber at a concentration of 0.5–1.5 x 106/mL. This unique, inert, immobilized bead mixture mimics the appearance of a 1:1 mixture of live and dead cells stained with trypan blue under brightfield illumination, which is the most common cell counting analysis method. The permanently mounted microspheres provide a stable sample that can be used repeatedly for verification of instrument performance. Please note that while the exact bead concentration for an individual slide is not specified, the bead concentration for each slide will fall in the 0.5–1.5 x 106/mL range and remain consistent over time.
Fluorescent microspheres enable fluorescence cell counting
The Countess 3 Standard Slide offers further utility by containing two populations of fluorescent beads that cover the fluorescence spectrum from blue to far red, thereby allowing analysis with a variety of EVOS light cube combinations available to Countess II FL and Countess 3 FL counter users. When viewed on the Countess 3 FL counter, the 15 μm microspheres are also fluorescent in a 1:1 mixture, providing approximately 50% detection with a DAPI or GFP light cube and approximately 50% detection with an RFP, Texas Red, or Cy5 light cube. The fluorescent microspheres enable users to perform or demonstrate fluorescence-based cell counting with the Countess 3 Standard Slide.
Stable standard for routine performance verification
The Countess 3 Standard Slide provides a stable sample containing microspheres that are visible in both brightfield and fluorescence channels and mimic a mixed population of live and dead cells. This allows users to easily perform routine verification of instrument performance for automated cell counting and demonstrate instrument usage. The immobilized microspheres eliminate sample variability, enabling reliable ongoing quality control.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dynamic Range0.5 to 1.5 million beads/mL
For Use With (Equipment)Countess II, II FL, Countess 3, and 3 FL
Light SourceBrightfield & Fluorescence
Quantity1 slide
Shipping ConditionAmbient
Warranty1 Year from date of purchase
WavelengthUV and Blue/Green and Red
Product LineCountess
Unit SizeEach
Contents & Storage
1 slide stored at ambient conditions.
Frequently asked questions (FAQs)
Where can I find the service retirement letter for the Countess II and II FL Automated Cell Counters?
You can find the service retirement letter for Countess II and II FL Automated Cell Counters below:
Important end-of-life communication for Countess II and II FL products
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Where can I find FAQs for Countess II and II FL Automated Cell Counters?
Countess II and II FL Automated Cell Counter FAQs can be found in our Product FAQ Database.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Where can I find a troubleshooting information for Countess 3 and 3 FL Automated Cell Counters?
Troubleshooting information for Countess 3 and 3FL can be found on the Cell Counting Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the shelf life for Countess Cell Counting Chamber Slides, disposable?
Our guarantee period for disposable Countess Cell Counting Chamber Slides is 5 years.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How can I find information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter?
For information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter, including 21 CFR Part 11, IQ/OQ, and warranty packages, please see refer to Countess Automated Cell Counter Services on our website.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.