Can I use OncoPro Tumoroid Culture Medium Kit to derive new tumoroid lines from patient-derived tissue such as tumor resections?
OncoPro Tumoroid Culture Medium Kit has been developed and optimized for the expansion and maintenance of existing tumoroid cultures. It has been used to successfully derive new tumoroid lines from patient tumor resections, dissociated tumor cells, and patient-derived xenografts, however the success rate of new derivations is highly variable and dependent on the quality of the starting tissue, tissue type, donor, etc.