VetMAX™ Fast Multiplex Mastermix, with ROX
Applied Biosystems™

VetMAX™ Fast Multiplex Mastermix, with ROX

VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for animal molecular diagnostics.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A57081100 Reactions
A57305500 Reactions
A573061000 Reactions
Catalog number A57081
Price (MXN)
3,716.56
Each
No. of Reactions:
100 Reactions
Price (MXN)
3,716.56
Each

VetMAX Fast Multiplex Master Mix with ROX is designed to be a fast-cycling-capable, multiplex-assay-compatible one-step RT-PCR master mix for testing in animal-derived samples. It can be used to amplify pathogenic DNA and RNA present in a variety of animal sample matrices, including ones with challenging PCR inhibitors. The fast-cycling capability, combined with multiplexing compatibility, allows for rapid and accurate real-time PCR results in <50 minutes.

Features of VetMAX Fast Multiplex Master Mix with ROX include:

  • Minimize time to results while maximizing accuracy and analytical sensitivity; amplify both DNA and RNA targets on the same plate using fast cycling
  • Improve flexibility by running up to four targets (not including the passive reference dye, ROX) on the same plate; run all assays using the same fast cycling thermal profile for improved efficiency
  • Increase analytical sensitivity of assays; formulated for increased analytical sensitivity while still being fast-cycling capable and multiplex-assay compatible
  • Strengthen reaction tolerance to inhibitors commonly found in animal-sourced sample matrices such as blood, feces, oral fluids, milk, and smegma

Fast and flexible

VetMAX Fast Multiplex Master Mix with ROX is ideal for fast-cycling, multiplex-capable molecular testing workflows. Due to its one-step RT-PCR capability, it can amplify both DNA and RNA targets using a single, fast-cycling thermal protocol that runs in <50 minutes.

Multiplex compatible

VetMAX Fast Multiplex Master Mix with ROX has been designed for multiplexing up to four targets (not including the passive reference channel, ROX). This meets the demand for more data per reaction and does so with increased analytical sensitivity and accuracy.

Robust and reliable

VetMAX Fast Multiplex Master Mix with ROX has been tested with animal-sourced samples of varying complexities ranging from simple samples like serum to complex samples like feces and porcine oral fluids. Its tolerance to PCR inhibition makes this product ideal for users who require workflow efficiency even when dealing with multiple sample types and pathogen assays.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Activation Time5 min.
CompatibilityTaqMan™
Final Product TypeMastermix
FormatTube
Hot StartBuilt-in Hot Start
No. of Reactions100 Reactions
PolymeraseAmpliTaq™ Fast DNA Polymerase
Quantity100 reactions
Starting MaterialDNA/RNA
Volume1 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Animal Health
FormLiquid
Label or DyeROX
PCR Method1-step RT-qPCR
Reaction SpeedFast
Unit SizeEach