E-Gel™ High-Throughput Mix Pack
Invitrogen™
E-Gel™ High-Throughput Mix Pack
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels.
Have Questions?
KatalognummerMenge
A573448 Gele
Katalognummer A57344
Preis (EUR)
337,00
Each
Menge:
8 Gele
Preis (EUR)
337,00
Each
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels. These gels use SYBR safe as a DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Benefits of using E-Gel agarose gels with SYBR Safe include:
  • Better sample integrity—blue-light excitation wavelength minimizes DNA damage, as opposed to UV light
  • Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with a separation time as short as 12 minutes
  • No buffers—E-Gel agarose gels do not require buffers to run
The box contains:
  • E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
  • E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
Running and imaging devices
High-throughput E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap Plus Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Nur für Forschungszwecke.
Specifications
Menge8 Gele
SterilitätNon-sterile
FormatPrecast Gel
GeltypE-Gel
Trennbereich100 to 2000 bp
TrennverfahrenElectrophoresis
Trennung vonDNA
Unit SizeEach