KingFisher™ Apex Dx 96 PCR AB Platinum Package
Thermo Scientific™
KingFisher™ Apex Dx 96 PCR AB Platinum Package
The Thermo Scientific KingFisher Apex Dx Purification Instrument enables extraction of nucleic acids, proteins, and cells and has a small,Weitere Informationen
Have Questions?
KatalognummerMenge
A590351 Packung
Katalognummer A59035
Preis (EUR)
-
Menge:
1 Packung
The Thermo Scientific KingFisher Apex Dx Purification Instrument enables extraction of nucleic acids, proteins, and cells and has a small, compact footprint, making it ideal for use in diagnostic and development workflows. With its intuitive software, automation capabilities, and revolutionary magnetic-particle separation technology, this instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
The instrument is US IVD and IVDR (EU 2017/746) compliant.
Compact, stand-alone Instrument
The instrument is compact and stand-alone, allowing you to easily set up extraction runs, control the instrument, and send run history from the instrument without the need for a computer.
Adaptable sample throughput
The instrument offers interchangeable 96- and 24-magnetic heads and heating blocks, providing flexibility to adjust the sample throughput as needed.
AB Platinum instrument service plan for warranty period
Our AB Platinum instrument service plan is our top tier total solution designed not only to optimize instrument performance and uptime, but also provide a seamless experience in clinical and IVD laboratories. Protect your instrument with the AB Platinum plan and raise your level of confidence in your instrument.
IQOQ services designed for regulatory compliance
We offer on-site installation qualification and operational qualification (IQ/OQ) services designed to help you comply with regulatory requirements. IQOQ is included with the installation, and OQ is recommended after moving the instrument, upgrading software or hardware, planned maintenance, or critical repairs.
Get started with SmartStart Orientation
SmartStart Orientation is included in the instrument purchase to help you get up and running quickly. This orientation, available on-site or remotely, provides an in-depth understanding of the instrument with engaging and interactive training led by an experienced Field Application Scientist (FAS).
Optional Diomni Enterprise Software for connectivity and laboratory ecosystem management
Diomni software enables remote browser-based applications for multiple users and instrument connectivity for the convenience of your laboratory ecosystem management.
More information about the KingFisher Apex Dx Purification Instrument ›
More information about Diomni Software ›
Please contact your Sales Representative for ordering information.
Für In-vitro-Diagnostik.
Specifications
Abmessungen79 x 56 x 46 cm
Display10 in. Touch Display
Zur Verwendung mit (Anwendung)Infectious Diseases, Oncology, Diagnostics
ProduktlinieKingFisher
Menge1 Packung
VersandbedingungRaumtemperatur
Gewicht (metrisch)56 kg
Fassungsvermögen94 Samples
Unit SizeEach
Inhalt und Lagerung
• 1 KingFisher Apex Dx instrument configured with 96 PCR Head, PCR Heating Block, and 96 Heating Block
• Quick Reference Guide
• 1 Installation/qualification service and support under the AB Platinum Service contract