VersiCap Mat, 96-well, flat cap strips
Thermo Scientific™
VersiCap Mat, 96-well, flat cap strips
The Thermo Scientific VersiCap Mats, Flat Cap Strips are unique, with each strip of eight caps being linked to eachRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. per Case
AB1815100100
AB181525
Catalog number AB1815100
Price (MXN)
-
No. per Case:
100
The Thermo Scientific VersiCap Mats, Flat Cap Strips are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Features
• Multiple cap strips can be applied at the same time when sealing PCR plates, thereby reducing setup time
• Remove the need to cut up adhesive seals
• Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
• PCR
Related Products
VersiPlate PCR Strip Tube Plate, 96-well, low profile
VersiPlate PCR Strip Tube Plate, 96-well, low profile, white
VersiCap Mat, 96-well, domed cap strips
VersiCap Mat, 96-well, flat cap strips
VersiCap Mat, 96-well, ultra clear cap strips
Specifications
FormatStrips of 8 Flat Caps
No. per Case100
Product TypePCR Plate Cap Strip
Quantity100 mats
Certified RNase/DNase-freeYes
ColorClear
MaterialPolypropylene
No. of Wells96
Plate Compatibility96-well Plate
Product LineVersiPlate
Temperature Range (Metric)-20°C to +120°C
Unit SizeEach