VersiCap Mat, 96-well, domed cap strips
Thermo Scientific™
VersiCap Mat, 96-well, domed cap strips
The Thermo Scientific VersiCap Mats, Domed Cap Strips are unique, with each strip of eight caps being linked to eachRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. per Case
AB181025
AB1810100100
Catalog number AB1810
Price (USD)
95.25
Each
No. per Case:
25
Request bulk or custom format
Price (USD)
95.25
Each
The Thermo Scientific VersiCap Mats, Domed Cap Strips are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Features
• Multiple cap strips can be applied at the same time when sealing PCR plates, thereby reducing setup time
• Remove the need to cut up adhesive seals
• Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
• PCR
Related Products
VersiPlate PCR Strip Tube Plate, 96-well, low profile
VersiPlate PCR Strip Tube Plate, 96-well, low profile, white
VersiCap Mat, 96-well, domed cap strips
VersiCap Mat, 96-well, flat cap strips
VersiCap Mat, 96-well, ultra clear cap strips
Specifications
FormatStrips of 8 Domed Caps
No. per Case25
Product TypePCR Plate Cap Strip
Quantity25 mats
Temperature Range (Metric)-20°C to +120°C
Certified RNase/DNase-freeYes
ColorClear
MaterialPolypropylene
No. of Wells96
Plate Compatibility96-well Plate
Product LineVersiPlate
Unit SizeEach