Gel Loading Buffer II (Denaturing PAGE)
Invitrogen™

Gel Loading Buffer II (Denaturing PAGE)

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM8546G1 mL
AM854710 mL
Catalog number AM8546G
Price (USD)
63.65
Online Exclusive
66.00
Save 2.35 (4%)
Each
Quantity:
1 mL
Price (USD)
63.65
Online Exclusive
66.00
Save 2.35 (4%)
Each

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue. Supplied in one 1.4 mL bottle. All Invitrogen™ Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferDrying Solutions
FormLiquid
Gel CompatibilityDenaturing Polyacrylamide Gels
Quantity1 mL
Shipping ConditionRoom Temperature
FormatBottle
Label or DyeXylene Cyanol
Product LineAmbion
Unit SizeEach
Contents & Storage
Store at –20°C.