Thermo Scientific Chemicals

2,6-Dichlor-3-Nitrotoluol, 99 %, Thermo Scientific Chemicals

Katalognummer: B20520.18
50 g, Each
Thermo Scientific Chemicals

2,6-Dichlor-3-Nitrotoluol, 99 %, Thermo Scientific Chemicals

Katalognummer: B20520.18
50 g, Each
Menge
Katalognummer: B20520.18
auch als B20520-18 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
29682-46-0
IUPAC Name
1,3-dichloro-2-methyl-4-nitrobenzene
Molecular Formula
C7H5Cl2NO2
InChI Key
WBNZUUIFTPNYRN-UHFFFAOYSA-N
SMILES
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
Form
Powder or lumps
Assay (GC)
≥98.5%
Appearance (Color)
Pale yellow
Melting Point (clear melt)
51.0-58.0?C

Beschreibung

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
2,6-Dichloro-3-Nitrotoluol, wird als wichtiger Rohstoff und Zwischenprodukt in der organischen Synthese, in Pharmazeutika, Agrochemikalien und Farbstoffen verwendet.

Löslichkeit
Unlöslich in Wasser.

Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Stabil unter empfohlenen Lagerbedingungen. Von starken Oxidationsmitteln fernhalten.
RUO – Nur für Forschungszwecke

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water