Thermo Scientific Chemicals

Cyclohexyl methyl ketone, 95%, Thermo Scientific Chemicals

Catalog number: B21000.14
25 g, Each
Thermo Scientific Chemicals

Cyclohexyl methyl ketone, 95%, Thermo Scientific Chemicals

Catalog number: B21000.14
25 g, Each
Quantity
Catalog number: B21000.14
also known as B21000-14
Price (USD)
Price: 147.00
Online price: 124.65
Your price:
Quantity
-

Chemical Identifiers

CAS
823-76-7
IUPAC Name
1-cyclohexylethan-1-one
Molecular Formula
C8H14O
InChI Key
RIFKADJTWUGDOV-UHFFFAOYSA-N
SMILES
CC(=O)C1CCCCC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4480-1.4540 @ 20?C
Assay (GC)
≥94.0%

Description

It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text