Thermo Scientific Chemicals

Nonafluorobutanesulfonyl fluoride, 90+%, Thermo Scientific Chemicals

Catalog number: B21322.14
25 g, Each
Thermo Scientific Chemicals

Nonafluorobutanesulfonyl fluoride, 90+%, Thermo Scientific Chemicals

Catalog number: B21322.14
25 g, Each
Quantity
Catalog number: B21322.14
also known as B21322-14
Price (USD)
Price: 86.50
Online price: 75.65
Your price:
Quantity
-

Chemical Identifiers

CAS
375-72-4
IUPAC Name
nonafluorobutane-1-sulfonyl fluoride
Molecular Formula
C4F10O2S
InChI Key
LUYQYZLEHLTPBH-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(F)(=O)=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
>90.0%
Refractive Index
<1.3000 @ 20?C

Description

Benzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.

Solubility
Hydrolyses in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, baese..
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text