Thermo Scientific Chemicals

3-Nitrophenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21540.14
25 g, Each
Thermo Scientific Chemicals

3-Nitrophenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21540.14
25 g, Each
Quantity
Catalog number: B21540.14
also known as B21540-14
Price (USD)
Price: 447.00
Online price: 386.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1877-73-2
IUPAC Name
2-(3-nitrophenyl)acetic acid
Molecular Formula
C8H7NO4
InChI Key
WUKHOVCMWXMOOA-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
Cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
115.0-121.0?C

Description

3-Nitrophenylacetic acid, is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophenylacetic acid, is a useful synthetic intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text