Thermo Scientific Chemicals

L-(-)-Xylose, 99%

Catalog number: B21622.22
100 g, Each
Thermo Scientific Chemicals

L-(-)-Xylose, 99%

Catalog number: B21622.22
100 g, Each
Quantity
Catalog number: B21622.22
also known as B21622-22
Price (USD)
Price: 503.00
Online price: 434.65
Your price:
Quantity
-

Chemical Identifiers

CAS
609-06-3
IUPAC Name
2,3,4,5-tetrahydroxypentanal
Molecular Formula
C5H10O5
InChI Key
PYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C=O
Optical Rotation
-18° to -20° (c=10 in water)
Assay from Suppliers CofA
≥98.5% (Unspecified)
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text