Thermo Scientific Chemicals

Sodium dichloroacetate, 96%, Thermo Scientific Chemicals

Catalog number: B21827.30
250 g, Each
Thermo Scientific Chemicals

Sodium dichloroacetate, 96%, Thermo Scientific Chemicals

Catalog number: B21827.30
250 g, Each
Quantity
Catalog number: B21827.30
also known as B21827-30
Price (USD)
Price: 1,010.00
Online price: 858.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2156-56-1
IUPAC Name
sodium 2,2-dichloroacetate
Molecular Formula
C2HCl2NaO2
InChI Key
LUPNKHXLFSSUGS-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C(Cl)Cl
Water Content (Karl Fischer Titration)
≤2.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Ethanol)
≥95.0 to ≤105.0%

Description

Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text