Polyethylenglycol 10,000, Thermo Scientific Chemicals
Polyethylenglycol 10,000, Thermo Scientific Chemicals
Polyethylenglycol 10,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylenglycol 10,000, Thermo Scientific Chemicals

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1000 g
Katalognummer B21955.0B
auch als B21955-0B bezeichnet
Preis (EUR)
69,65
Exklusiv online
77,40
Ersparnis 7,75 (10%)
Each
Menge:
1000 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
69,65
Exklusiv online
77,40
Ersparnis 7,75 (10%)
Each
Chemikalien-Kennzeichnungen
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetDatenblatt
Solution TestClear, APHA ≤30
Viscosity550 - 750 mPas (50%, water @ 20?C)
Appearance (Color)White
FormPowder or crystals or flakes
CommentAveraged mol weight 9000 - 11250 (based on OH value)
Mehr anzeigen
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only