Thermo Scientific Chemicals

Cesium hydroxide monohydrate, 96%; cesium carbonate <5%, Thermo Scientific Chemicals

Catalog number: B22480.36
500 g, Each
Thermo Scientific Chemicals

Cesium hydroxide monohydrate, 96%; cesium carbonate <5%, Thermo Scientific Chemicals

Catalog number: B22480.36
500 g, Each
Quantity
Catalog number: B22480.36
also known as B22480-36
Price (USD)
Price: 732.00
Online price: 622.65
Your price:
Quantity
-

Chemical Identifiers

CAS
35103-79-8
IUPAC Name
caesium(1+) hydrate hydroxide
Molecular Formula
CsH3O2
InChI Key
ABSOMGPQFXJESQ-UHFFFAOYSA-M
SMILES
O.[OH-].[Cs+]
Appearance (Color)
White
Form
Crystalline powder or powder
Assay (unspecified)
≥95.0% as monohydrate
Impurity content
Cesium carbonate ≤5.0%
Trace Metal
≤0.05%

Description

Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

Solubility
Soluble in water with exothermic reaction.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, acids, air, water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text