3-Amino-9-ethylcarbazol, 95 %, Thermo Scientific Chemicals
3-Amino-9-ethylcarbazol, 95 %, Thermo Scientific Chemicals
3-Amino-9-ethylcarbazol, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-9-ethylcarbazol, 95 %, Thermo Scientific Chemicals

CAS: 132-32-1 | C14H14N2 | 210.28 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer B22529.14
auch als B22529-14 bezeichnet
Preis (EUR)
70,65
Exklusiv online
78,30
Ersparnis 7,65 (10%)
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
70,65
Exklusiv online
78,30
Ersparnis 7,65 (10%)
Each
Chemikalien-Kennzeichnungen
CAS132-32-1
IUPAC Name9-ethyl-9H-carbazol-3-amine
Molecular FormulaC14H14N2
InChI KeyOXEUETBFKVCRNP-UHFFFAOYSA-N
SMILESCCN1C2=C(C=CC=C2)C2=C1C=CC(N)=C2
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Proton NMRConforms to structure (U.S. specification)
Appearance (Color)Yellow to brown
Assay (GC)≥94.0% (non-U.S. specification)
Assay from Suppliers CofA≥94.0% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Mehr anzeigen
Peroxidase substrate for immunoblotting and immunohistochemical staining procedures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only