2-Fluoralphamethyl-4-Biphenylessigsäure, 99 %, Thermo Scientific Chemicals
2-Fluoralphamethyl-4-Biphenylessigsäure, 99 %, Thermo Scientific Chemicals
2-Fluoralphamethyl-4-Biphenylessigsäure, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoralphamethyl-4-Biphenylessigsäure, 99 %, Thermo Scientific Chemicals

CAS: 5104-49-4 | C15H13FO2 | 244.27 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
Katalognummer B22603.03
auch als B22603-03 bezeichnet
Preis (EUR)
74,10
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
74,10
Each
Chemikalien-Kennzeichnungen
CAS5104-49-4
IUPAC Name2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Molecular FormulaC15H13FO2
InChI KeySYTBZMRGLBWNTM-UHFFFAOYNA-N
SMILESCC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Melting Point (clear melt)111.5-115.5?C
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
FormPowder
A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only