Thermo Scientific Chemicals

Citronellic acid, 94%, Thermo Scientific Chemicals

Catalog number: B22862.14
25 g, Each
Thermo Scientific Chemicals

Citronellic acid, 94%, Thermo Scientific Chemicals

Catalog number: B22862.14
25 g, Each
Quantity
Catalog number: B22862.14
also known as B22862-14
Price (USD)
Price: 75.10
Online price: 63.65
Your price:
Quantity
-

Chemical Identifiers

CAS
502-47-6
IUPAC Name
3,7-dimethyloct-6-enoic acid
Molecular Formula
C10H18O2
InChI Key
GJWSUKYXUMVMGX-UHFFFAOYNA-N
SMILES
CC(CCC=C(C)C)CC(O)=O
Assay (Silylated GC)
≥92.0% (sum of isomers)
Form
Liquid
Refractive Index
1.4510-1.4550 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text