Thermo Scientific Chemicals

3-Methylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23025.06
5 g, Each
Thermo Scientific Chemicals

3-Methylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23025.06
5 g, Each
Quantity
Catalog number: B23025.06
also known as B23025-06
Price (USD)
Price: 113.00
Online price: 98.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17933-03-8
IUPAC Name
(3-methylphenyl)boronic acid
Molecular Formula
C7H9BO2
InChI Key
BJQCPCFFYBKRLM-UHFFFAOYSA-N
SMILES
CC1=CC=CC(=C1)B(O)O
Assay (HPLC)
≥96.0%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
152.0-162.0?C

Description

Suzuki coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text