Thermo Scientific Chemicals

5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: B24018.22
100 g, Each
Thermo Scientific Chemicals

5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: B24018.22
100 g, Each
Quantity
Catalog number: B24018.22
also known as B24018-22
Price (USD)
Price: 62.80
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15454-54-3
IUPAC Name
2H-1,2,3,4-tetrazol-5-amine hydrate
Molecular Formula
CH5N5O
InChI Key
JVSMPWHQUPKRNV-UHFFFAOYSA-N
SMILES
O.NC1=NNN=N1
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or crystalline powder or powder
Water Content (Karl Fischer Titration)
16.0-18.9% (0.9 to 1.1 waters)
Appearance (Color)
White

Description

5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

Solubility
Soluble in water, ethanol (1.36%).

Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text