Thermo Scientific Chemicals

N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals

Catalog number: B24529.22
100 g, Each
Thermo Scientific Chemicals

N,N-Dimethylthioformamide, 97%, Thermo Scientific Chemicals

Catalog number: B24529.22
100 g, Each
Quantity
Catalog number: B24529.22
also known as B24529-22
Price (USD)
Price: 528.00
Online price: 451.65
Your price:
Quantity
-

Chemical Identifiers

CAS
758-16-7
IUPAC Name
N,N-dimethylmethanethioamide
Molecular Formula
C3H7NS
InChI Key
SKECXRFZFFAANN-UHFFFAOYSA-N
SMILES
CN(C)C=S
Appearance (Color)
Clear, colorless to yellow or pale orange
Form
Liquid
Assay (GC)
≥96.0%

Description

It is used in telecommunication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in telecommunication.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text