Thermo Scientific Chemicals

Methyl valerate, 99%, Thermo Scientific Chemicals

Catalog number: B24828.36
500 g, Each
Thermo Scientific Chemicals

Methyl valerate, 99%, Thermo Scientific Chemicals

Catalog number: B24828.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B24828.36
also known as B24828-36
Price (USD)
Price: 108.00
Online price: 91.65
Your price:
Quantity
-

Chemical Identifiers

CAS
624-24-8
IUPAC Name
methyl pentanoate
Molecular Formula
C6H12O2
InChI Key
HNBDRPTVWVGKBR-UHFFFAOYSA-N
SMILES
CCCCC(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Free acid (titration)
≤0.5%
Refractive Index
1.3965-1.3995 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text