Thermo Scientific Chemicals

m-Xylylene dibromide, 97%, Thermo Scientific Chemicals

Catalog number: B24959.18
50 g, Each
Thermo Scientific Chemicals

m-Xylylene dibromide, 97%, Thermo Scientific Chemicals

Catalog number: B24959.18
50 g, Each
Quantity
Catalog number: B24959.18
also known as B24959-18
Price (USD)
Price: 274.00
Online price: 234.65
Your price:
Quantity
-

Chemical Identifiers

CAS
626-15-3
IUPAC Name
1,3-bis(bromomethyl)benzene
Molecular Formula
C8H8Br2
InChI Key
OXHOPZLBSSTTBU-UHFFFAOYSA-N
SMILES
BrCC1=CC(CBr)=CC=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
71.0-81.0?C

Description

It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, active metals, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text