Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: B25123.03
1 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: B25123.03
1 g, Each
Quantity
Catalog number: B25123.03
also known as B25123-03
Price (USD)
Price: 45.10
Online price: 38.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2233-18-3
IUPAC Name
4-hydroxy-3,5-dimethylbenzaldehyde
Molecular Formula
C9H10O2
InChI Key
UYGBSRJODQHNLQ-UHFFFAOYSA-N
SMILES
CC1=CC(C=O)=CC(C)=C1O
Form
Crystals or Crystalline powder
Assay (GC)
≥97.0%
Melting Point (clear melt)
111.5-117.5?C
Appearance (Color)
White to cream or yellow to orange or pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text