Thermo Scientific Chemicals

Dimethyl acetylsuccinate, 96%, Thermo Scientific Chemicals

Catalog number: B25423.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl acetylsuccinate, 96%, Thermo Scientific Chemicals

Catalog number: B25423.36
500 g, Each
Quantity
Catalog number: B25423.36
also known as B25423-36
Price (USD)
Price: 138.00
Online price: 117.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10420-33-4
IUPAC Name
1,4-dimethyl 2-acetylbutanedioate
Molecular Formula
C8H12O5
InChI Key
XREKLQOUFWBSFH-UHFFFAOYNA-N
SMILES
COC(=O)CC(C(C)=O)C(=O)OC
Form
Fused solid
Melting Point (clear melt)
28.0-37.0?C
Appearance (Color)
White
Assay (GC)
≥95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text