Thermo Scientific Chemicals

3-Formylbenzeneboronic acid, Thermo Scientific Chemicals

Catalog number: B25437.14
25 g, Each
Thermo Scientific Chemicals

3-Formylbenzeneboronic acid, Thermo Scientific Chemicals

Catalog number: B25437.14
25 g, Each
Quantity
Catalog number: B25437.14
also known as B25437-14
Price (USD)
Price: 338.00
Online price: 292.65
Your price:
Quantity
-

Chemical Identifiers

CAS
87199-16-4
IUPAC Name
(3-formylphenyl)boronic acid
Molecular Formula
C7H7BO3
InChI Key
HJBGZJMKTOMQRR-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC(C=O)=C1
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale orange to pale brown
Form
Crystals or powder or crystalline powder or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text