How are the Escherichia coli (K-12 strain) BioParticles, fluorescein conjugate supplied?
The Escherichia coli (K-12 strain) BioParticles, fluorescein conjugate are supplied as lyophilized powder in a 10 mg.unit size. There are approximately 3 x 10E8 E. coli particles per mg solid.