ProcartaPlex™ Human Cytokine/Chemokine/Growth Factor Panel 1, 45plex
Invitrogen™

ProcartaPlex™ Human Cytokine/Chemokine/Growth Factor Panel 1, 45plex

The ProcartaPlex Human Cytokine/Chemokine/Growth Factor Panel 1 45plex enables the exploration of immune function by analyzing 45 protein targets in深入閱讀
Have Questions?
產品號碼Quantity
EPX450-12171-90196 Tests
產品號碼 EPX450-12171-901
價格 (HKD)
70,703.00
Each
Quantity:
96 Tests
價格 (HKD)
70,703.00
Each

The ProcartaPlex Human Cytokine/Chemokine/Growth Factor Panel 1 45plex enables the exploration of immune function by analyzing 45 protein targets in a single well using Luminex xMAP technology. The panel is comprised of five modular sub-panels that allow for an initial broad screen to determine targets of interest and subsequent studies analyzing specific functional subsets of cytokines that can be purchased separately. The sub-panels are fully combinable with each other or with individual simplex assays corresponding to the targets in the panel. The same targets can be found in the Human Cytokine/Chemokine/Growth Factor Convenience Panel 1 45plex (Cat. No. EPXR450-12171-901) that is in a ready-to-use format that requires less pipetting.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit our ProcartaPlex Immunoassays page for more information and available products.

Target list [bead region]:
Th1/Th2: GM-CSF [44], IFN gamma [43], IL-1 beta [18], IL-2 [19], IL-4 [20], IL-5 [21], IL-6 [25], IL-8 [27], IL-12p70 [34], IL-13 [35], IL-18 [66], TNF alpha [45]

Th9/Th17/Th22/Treg: IL-9 [52], IL-10 [28], IL-17A (CTLA-8) [36], IL-21 [72], IL-22 [76], IL-23 [63], IL-27 [14]

Inflammatory cytokines: IFN alpha [48], IL-1 alpha [62], IL-1RA [38], IL-7 [26], IL-15 [65], IL-31 [37], LIF [15], TNF beta [54]

Chemokines. Eotaxin (CCL11) [33], GRO alpha (CXCL1) [61], IP-10 (CXCL10) [22], MCP-1 (CCL2) [51], MIP-1 alpha (CCL3) [12], MIP-1 beta (CCL4) [47], RANTES (CCL5) [42], SDF-1 alpha [13]

Growth factors: BDNF [57], EGF [56], FGF-2 [75], HGF [46], NGF beta [55], PDGF-BB [77], PlGF-1 [29], SCF [39], VEGF-A [78], VEGF-D [53]

About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:
• Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
• More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
• Well-established Luminex technology highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

For Research Use Only. Not for use in diagnostic procedures.
規格
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
For Use With (Equipment)Luminex™ Instruments
FormatMultiplex Kit
Product LineProcartaPlex
Sample TypeSerum, Plasma, Cell Culture Supernatants
Sample VolumeSerum, Plasma: 25 μL; CCS: 50 μL
Shipping ConditionWet Ice
CombinabilityCombinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaImmunology, Cytokines, Chemokines, Growth Factors
SpeciesHuman
Unit SizeEach
內容物與存放

• 2 vials Human Standard Mix A (lyophilized)
• 2 vials Human Standard Mix B (lyophilized)
• 2 vials Human Standard Mix C (lyophilized)
• 1 vial Capture Bead Mix B (1X)
• 1 vial Capture Bead Mix C (1X)
• 1 vial Capture Bead Mix D (1X)
• 1 vial Capture Bead Mix F (1X)
• 1 vial Capture Bead Mix G (1X)
• 1 vial Biotinylated Detection Antibody Mix B (50X)
• 1 vial Biotinylated Detection Antibody Mix C (50X)
• 1 vial Biotinylated Detection Antibody Mix D (50X)
• 1 vial Biotinylated Detection Antibody Mix F (50X)
• 1 vial Biotinylated Detection Antibody Mix G (50X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Streptavidin-PE (1X)

常見問答集 (常見問題)

What is the size of the Luminex beads you currently use?

The beads used in our Luminex instrument-compatible ProcartaPlex and QuantiGene Plex assays are 6.5 micron superparamagnetic beads.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am interested in performing Luminex assays using BioSource kits, and I have a Luminex xMAP system. Besides the kits and system, what other reagents and equipment will I need?

The following is a list of general lab supplies that are required for running BioSource immunoassays on the Luminex xMAP system:
1) Sonicating water bath
2) Orbital shaker
3) Vortexer
4) Repeating and/or multi-channel pipetter (not required, but recommended)
5) Calibrated adjustable precision pipettes, with disposable plastic tips
6) Glass/plastic tubes and racks for preparing reagents
7) Graduated cylinder and container for preparing wash solution
8) Aluminum foil
9) Deionized or distilled water.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do the Luminex beads require special care in handling?

The Luminex beads should be protected from light because they are susceptible to photobleaching. We recommend protecting the beads by keeping containers covered with aluminum foil during all incubation steps, and exercising care during handling. The beads should not be frozen, subjected to excessive heat, or exposed to organic solvents.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why would the Luminex acquisition software display "Sample Empty" messages during analysis?

(1) The user did not properly aliquot the diluted beads, such that no beads were actually added to the wells (make sure that the bead concentrates are sonicated and vortexed well, then check the pipet tip to ensure that air bubbles were not drawn up)
(2) The user missed loading diluted beads to some wells, which is likely since the small volume is clear and difficult to visualize in the clear plastic plate (we have now addressed this customer difficulty by coloring each of the Buffer Reagent Kit components)
(3) The user applied too much vacuum pressure at some point during the wash steps, or allowed the pressure to spike even once, such that the filter membrane tore in a few wells releasing the beads (make sure that the vacuum manifold pressure is kept below 5mm/in Hg, depending on their system -- a good rule of thumb is that it should take a full 3-second count to GENTLY empty the wells of 200uL)
(4) The user did not properly sonicate and vortex the beads prior to dilution, such that the percent of bead aggregation was high and the instrument was unable to find enough single beads to meet the events/bead value designated by the customer (make sure that the Bead Concentrate tube is put into the waterbath all the way to the cap, since the tube is hollow until the top third)
(5) The user lost beads by shaking the plate too aggressively or handling it improperly (make sure that the orbital shaker is set to a speed that allows for maximum vortex in the wells without spillage)
(6) The user exposed the beads to an excess of light during storage or running of the assay, such that some but not all of the beads were photobleached and therefore falling outside the acceptable range for each bead region (make sure that the plate is covered on the top/sides with foil throughout the assay, away from Windows and spotlights, and that the bead component of the kits is stored in the dark)*
(7) There was a clog in the sample needle, such that the instrument was unable to take up enough sample to meet the number of events requested per bead region (suggest that the user follow the manual instructions for dislodging a clog, which include several Back Flush steps and may require removal of the needle for sonication with probe alignment).

* Some of the older Antibody Bead Kits still have clear plastic tops instead of black ones. In cases where customers store kits in lit refrigerators, or keep them open on the lab bench, even a few hours of light exposure is enough to photobleach beads. It is important to note, in general, that higher number bead regions are more susceptible to photobleaching. In order to draw conclusions about the source of the difficulty, we would ask to see the data, specifically the Masterplex QT file, which would enable us to examine the pattern of "Sample Empty" occurrences in addition to the bead counts per well.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the Luminex beads made of?

The beads are made of polystyrene.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

引用資料與參考文獻 (25)

引用資料與參考文獻
Abstract
Decreased memory B cell frequencies in COVID-19 delta variant vaccine breakthrough infection.
Authors:Tay MZ, Rouers A, Fong SW, Goh YS, Chan YH, Chang ZW, Xu W, Tan CW, Chia WN, Torres-Ruesta A, Amrun SN, Huang Y, Hor PX, Loh CY, Yeo NK, Wang B, Ngoh EZX, Salleh SNM, Chavatte JM, Lim AJ, Maurer-Stroh S, Wang LF, Lin RVTP, Wang CI, Tan SY, Young BE, Leo YS, Lye DC, Renia L, Ng LF
Journal:EMBO Mol Med
PubMed ID:34994081
The SARS-CoV-2 Delta (B.1.617.2) variant is capable of infecting vaccinated persons. An open question remains as to whether deficiencies in specific vaccine-elicited immune responses result in susceptibility to vaccine breakthrough infection. We investigated 55 vaccine breakthrough infection cases (mostly Delta) in Singapore, comparing them against 86 vaccinated close contacts who ... More
Effects of altered salt intake and diet on cytokines in humans: A 20-week randomized cross-over intervention study.
Authors:Niiranen T,Erlund I,Jalkanen S,Jula A,Salmi M
Journal:European journal of immunology
PubMed ID:36330564
Zika Virus Infection Preferentially Counterbalances Human Peripheral Monocyte and/or NK Cell Activity.
Authors:Lum FM, Lee D, Chua TK, Tan JJL, Lee CYP, Liu X, Fang Y, Lee B, Yee WX, Rickett NY, Chia PY, Lim V, Leo YS, Matthews DA, Hiscox JA, Ng LFP
Journal:
PubMed ID:29600283
'Zika virus (ZIKV) has reemerged in the population and caused unprecedented global outbreaks. Here, the transcriptomic consequences of ZIKV infection were studied systematically first in human peripheral blood CD14' ... More
Influenza Virus Infection Enhances Antibody-Mediated NK Cell Functions via Type I Interferon-Dependent Pathways.
Authors:Jegaskanda S, Vanderven HA, Tan HX, Alcantara S, Wragg KM, Parsons MS, Chung AW, Juno JA, Kent SJ
Journal:J Virol
PubMed ID:30541850
'Natural killer (NK) cells are an important component in the control of influenza virus infection, acting to both clear virus-infected cells and release antiviral cytokines. Engagement of CD16 on NK cells by antibody-coated influenza virus-infected cells results in antibody-dependent cellular cytotoxicity (ADCC). Increasing the potency of antibody-mediated NK cell activity ... More
Deciphering the state of immune silence in fatal COVID-19 patients.
Authors:Bost P, De Sanctis F, Canè S, Ugel S, Donadello K, Castellucci M, Eyal D, Fiore A, Anselmi C, Barouni RM, Trovato R, Caligola S, Lamolinara A, Iezzi M, Facciotti F, Mazzariol A, Gibellini D, De Nardo P, Tacconelli E, Gottin L, Polati E, Schwikowski B, Amit I, Bronte V
Journal:Nat Commun
PubMed ID:33674591
'Since the beginning of the SARS-CoV-2 pandemic, COVID-19 appeared as a unique disease with unconventional tissue and systemic immune features. Here we show a COVID-19 immune signature associated with severity by integrating single-cell RNA-seq analysis from blood samples and broncho-alveolar lavage fluids with clinical, immunological and functional ex vivo data. This ... More