IC Fixation Buffer - FAQs
IC Fixation Buffer - FAQs
View additional product information for IC Fixation Buffer - FAQs (FB001)
1 product FAQs found
What fixative should I use to fix my cells or tissues for antibody labeling?
Aldehyde-based fixatives (e.g., formaldehyde, glutaraldehyde) crosslink various cellular components, which helps retain proteins and cell morphology, but some antigens can be masked by the crosslinking, requiring antigen retrieval methods to unmask the antigen binding sites. Also, aldehyde-based fixation requires permeabilization to allow entry of antibodies. Organic solvents such as methanol and acetone condense proteins and permeabilize the cells, but cell morphology can be compromised. Surface antigens can be labeled without fixation.