E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2%
Invitrogen™
E-Gel™ Power Snap Electrophoresis System Starter Kit, DC Safe 2%
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, DC Safe 2%, provides everything you need to quickly start usingRead more
Have Questions?
Catalog NumberQuantity
G8372ST1 kit
Catalog number G8372ST
Price (MXN)
-
Quantity:
1 kit
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, DC Safe 2%, provides everything you need to quickly start using precast 1% E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
  • Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)
Learn more about the E-Gel Power Snap Electrophoresis System ›
Learn more about precast E-Gel EX agarose gel technology ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Gel SizeMini
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)
Store at room temperature.