(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
(S)-(-)-3-Amino-1,2-Propandiol wird als chiraler Selektorligand verwendet, der anschließend verwendet wird für eine nucleophile Substitution mit dem Dimethoxytrityl (DMT)-geschützten
Löslichkeit
Wasserlöslich.
Hinweise
Hygroskopisch. Behälter dicht verschlossen lassen. Kühl und trocken in einem gut versiegelten Behälter lagern.
RUO – Research Use Only
Allgemeine Referenzen:
- Guijun Wang.; Rawle I. Hollingsworth. Direct Conversion of (S)-3-Hydroxy-γ-butyrolactone to Chiral Three-Carbon Building Blocks. J. Org. Chem. 1999, 64, (3), 1036-1038.
- Pham Thi Thanh Ha.; Jos Hoogmartens.; Ann Van Schepdael. Recent advances in pharmaceutical applications of chiral capillary electrophoresis. Journal of Pharmaceutical and Biomedical Analysis. 2006, 41, (1), 1-11.