cis-4-Hydroxy-L-Prolin, 99 %, Thermo Scientific Chemicals
cis-4-Hydroxy-L-Prolin, 99 %, Thermo Scientific Chemicals
cis-4-Hydroxy-L-Prolin, 99 %, Thermo Scientific Chemicals
cis-4-Hydroxy-L-Prolin, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-4-Hydroxy-L-Prolin, 99 %, Thermo Scientific Chemicals

CAS: 618-27-9 | C5H9NO3 | 131.13 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 mg
1 g
Katalognummer H27194.MD
auch als H27194-MD bezeichnet
Preis (EUR)
222,00
Each
Zum Warenkorb hinzufügen
Menge:
250 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
222,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS618-27-9
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to cream or pale yellow
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Optical Rotation-69.0° to -58° (c=2 in water)
cis-4-Hydroxy-L-proline (CHP) is a proline analog that inhibits collagen synthesis and has been used as an anticancer compound. CHP blocked myotube formation and expression of sarcomeric myosin heavy chain in C2C12 murine skeletal muscle cells. CHP inhibited proliferation of murine Panc02 pancreatic carcinoma cell line3 and rat pancreatic carcinoma cell line DSL6A. cis-4-Hydroxy-L-proline and other hydrosyl-L-prolines are important intermediates for chiral synthesis of potential drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Wasserlöslich.

Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von starken Oxidationsmitteln fernhalten.
RUO – Research Use Only

Allgemeine Referenzen:

  1. Heather Dickens et. al. Anticancer drug cis-4-hydroxy-L-proline: Correlation of preclinical toxicology with clinical parameters of liver function. Molecular Medicine Reports. 2008, 1(4), 459-464 .
  2. Roux A, Xu Y,; Heilier JF,; Olivier MF,; Ezan E,; Tabet JC,; Junot C. Annotation of the human adult urinary metabolome and metabolite identification using ultra high performance liquid chromatography coupled to a linear quadrupole ion trap-Orbitrap mass spectrometer. Analytische Chemie. 2012, 84, 6429-6437.