Thermo Scientific Chemicals

4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals

Catalog number: H27330.06
5 g, Each
Thermo Scientific Chemicals

4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals

Catalog number: H27330.06
5 g, Each
Quantity
Catalog number: H27330.06
also known as H27330-06
Price (USD)
Price: 650.00
Online price: 557.65
Your price:
Quantity
-

Chemical Identifiers

CAS
261733-18-0
IUPAC Name
4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Molecular Formula
C36H27NOP2
InChI Key
HSWZLYXRAOXOLL-UHFFFAOYSA-N
SMILES
N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream or pale yellow or pale green or green/grey
Form
Crystals or powder or crystalline powder
Assay by NMR
≥98.0%
Identification (FTIR)
Conforms
Melting Point
243-259?C (may decompose).

Description

4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text