Thermo Scientific Chemicals

4-Cyanopiperidine, 96%, Thermo Scientific Chemicals

Catalog number: H27722.14
25 g, Each
Thermo Scientific Chemicals

4-Cyanopiperidine, 96%, Thermo Scientific Chemicals

Catalog number: H27722.14
25 g, Each
Quantity
Catalog number: H27722.14
also known as H27722-14
Price (USD)
Price: 225.00
Your price:
Quantity
-

Chemical Identifiers

CAS
4395-98-6
IUPAC Name
piperidine-4-carbonitrile
Molecular Formula
C6H10N2
InChI Key
FSDNTQSJGHSJBG-UHFFFAOYSA-N
SMILES
N#CC1CCNCC1
Refractive Index
1.4725-1.4785 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless to pale yellow

Description

4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text