It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Es ist ein wichtiger Rohstoff und Zwischenprodukt, der in der organischen Synthese, Pharmazeutika, Agrochemikalien und Farbstoffen verwendet wird.
Löslichkeit
Schwer in Wasser löslich.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von Oxidationsmitteln entfernt aufbewahren.
RUO – Research Use Only
Allgemeine Referenzen:
- Mårten Jacobsson; Ulf Ellervik; Mattias Belting; Katrin Mani. Selective antiproliferative activity of hydroxynaphthyl-beta-D-xylosides. Journal of Medicinal Chemistry. 2006, 49(6), 1932-1938.
- Cheng-Lin Chung; Wen-Fu Lee; Chun-Hung Lin; Sheng-Huei Hsiao.Journal of Polymer Science Part A: Polymer Chemistry. Journal of Medicinal Chemistry. 2009 , 47(7), 1756-1770.