Thermo Scientific Chemicals

Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H29107.03
1 g, Each
Thermo Scientific Chemicals

Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H29107.03
1 g, Each
Quantity
Catalog number: H29107.03
also known as H29107-03
Price (USD)
Price: 47.10
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
139631-62-2
IUPAC Name
cyclopropanesulfonyl chloride
Molecular Formula
C3H5ClO2S
InChI Key
PFWWSGFPICCWGU-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1CC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.4740-1.4790 @ 20?C
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text