Thermo Scientific Chemicals

Methyl isocyanoacetate, 95%, Thermo Scientific Chemicals

Catalog number: H31069.06
5 g, Each
Thermo Scientific Chemicals

Methyl isocyanoacetate, 95%, Thermo Scientific Chemicals

Catalog number: H31069.06
5 g, Each
Quantity
Catalog number: H31069.06
also known as H31069-06
Price (USD)
Price: 107.00
Your price:
Quantity
-

Chemical Identifiers

CAS
39687-95-1
IUPAC Name
methyl 2-(methylideneamino)acetate
Molecular Formula
C4H7NO2
InChI Key
ZRGVTTCCASYOFN-UHFFFAOYSA-N
SMILES
COC(=O)CN=C
Appearance (Color)
Clear pale yellow to dark brown
Form
liquid
Assay (GC)
> 94.0%
Refractive Index
1.4140 - 1.4200 @20?C

Description

Methyl isocyanoacetate is used in the copper-catalyzed, diastereoselective synthesis of oxazolines. It is involved in the direct aldol reaction with carbonyl compounds to prepare corresponding oxazoline by using catechol-copper network catalyst. Further, it is also involved in four-component Ugi condensation reaction. It undergoes asymmetric aldol reaction with fluorinated benzaldehyde using gold(I) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl isocyanoacetate is used in the copper-catalyzed, diastereoselective synthesis of oxazolines. It is involved in the direct aldol reaction with carbonyl compounds to prepare corresponding oxazoline by using catechol-copper network catalyst. Further, it is also involved in four-component Ugi condensation reaction. It undergoes asymmetric aldol reaction with fluorinated benzaldehyde using gold(I) as a catalyst.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong acids, hydrated iron(II) sulfate, heavy metals and aluminum. Store in a cool place. Keep the container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text