Stains-All, 96 %
Stains-All, 96 %
Stains-All, 96 %
Stains-All, 96 %
Thermo Scientific Chemicals

Stains-All, 96 %

CAS: 7423-31-6 | C30H27BrN2S2
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5g
Katalognummer H32127.06
auch als H32127-06 bezeichnet
Preis (EUR)
281,00
Each
Menge:
5g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
281,00
Each
Chemikalien-Kennzeichnungen
CAS7423-31-6
IUPAC Name1-ethyl-2-{3-[(2Z)-1-ethyl-1H,2H-naphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Molecular FormulaC30H27BrN2S2
InChI KeyMPBRYMWMMKKRGC-UHFFFAOYSA-M
SMILES[Br-].CCN1\C(SC2=CC=C3C=CC=CC3=C12)=C\C(C)=CC1=[N+](CC)C2=C(S1)C=CC1=CC=CC=C21
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Dark blue or dark grey to black
FormPowder
Assay (Dye)≥95.0%
Elemental AnalysisCarbon, C: 61.17-67.61% (Theory: 64.39%)
Elemental AnalysisNitrogen, N: 4.75-5.25% (Theory: 5.00%)
General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Häufig gestellte Fragen (FAQ)

Is Stains-All, 96% suitable for RNA purification by SDS-PAGE?

Yes, Stains-All, 96% is a cationic cyanine dye, and it is possible to stain RNA with it. Gel staining should be performed in the dark, as cyanine dyes can be excited with visible light to produce ROS, leading to damage of biomolecules.