5-Bromooxindole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
5-Bromoxindol ist ein wichtiger Rohstoff und ein wichtiges Zwischenprodukt für organische Synthesen, Pharmazeutika, Agrochemikalien und Farbstoffe.
Löslichkeit
Löslich in Dimethylformamide.
Hinweise
In einem kühlen, trockenen, gut belüfteten Bereich lagern, fernab von unverträglichen Substanzen wie Oxidationsmitteln.
RUO – Research Use Only
Yogesh S. Tingre; Ming-Tai Shen; Chaochin Su; Shih-Yu Ho; Sheng-Han Tsai; Bo-Ren Chen und Wen-Ren Li. Novel Oxindole Based Sensitizers: Synthesis and Application in Dye-Sensitized Solar Cells. Org. Lett.2013, 15, (17), 4292-4295.