N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
N-Fmoc-3-(2-Naphthyl)-L-Alanin wird als Zwischenprodukt der organischen chemischen Synthese verwendet.
Löslichkeit
Schwer in Wasser löslich.
Hinweise
Stabil unter empfohlenen Lagerbedingungen. Von Oxidationsmitteln entfernt aufbewahren
RUO – Research Use Only