Thermo Scientific Chemicals
5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Catalog number: H52330.MD
250 mg, Each
Thermo Scientific Chemicals
5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Catalog number: H52330.MD
250 mg, Each
Quantity
Catalog number: H52330.MD
also known as H52330-MD
Price (USD)
Quantity
-
Description
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.