Thermo Scientific Chemicals

N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: H52795.06
5 g, Each
Thermo Scientific Chemicals

N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: H52795.06
5 g, Each
Quantity
Catalog number: H52795.06
also known as H52795-06
Price (USD)
Price: 1,302.00
Online price: 1,106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106627-54-7
IUPAC Name
sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
Molecular Formula
C4H4NNaO6S
InChI Key
RPENMORRBUTCPR-UHFFFAOYNA-M
SMILES
[Na+].ON1C(=O)CC(C1=O)S([O-])(=O)=O
Assay (unspecified)
≥96.0%
Appearance (Color)
White to Pale yellow
Residual water
≤1%
Form
Powder

Description

N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

Solubility
Soluble in methanol and water.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text